Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHA7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548413

Catalog No. PA548413


Add to cart

Description

Description

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintece of specific neuronal connections in the brain.
Specifications

Specifications

PCDHA7
Polyclonal
PBS with 0.09% sodium azide
Q9UN72
Protocadherin alpha-7, PCDH-alpha-7, PCDHA7, CNRS4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56141
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PCDHA7
Liquid
PCDHA7
KLH conjugated synthetic peptide between 272-300 amino acids from the Central region of human PCDHA7.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit