Learn More
Aviva Systems Biology Corp PCDHAC1 Antibody - N-terminal region (ARP49341_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHAC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCDHAC1. Alternate names: PCDH-ALPHA-C1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60067