Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHB15 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548325

Catalog No. PA548325

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintece of specific neuronal connections in the brain.
Specifications

Specifications

PCDHB15
Polyclonal
PBS with 0.09% sodium azide
Q9Y5E8
Protocadherin beta-15, PCDH-beta-15, PCDHB15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56121
Western Blot
Unconjugated
PCDHB15
Liquid
PCDHB15
KLH conjugated synthetic peptide between 113-140 amino acids from the N-terminal region of human PCDHB15.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit