Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$143.00 - $405.00
Specifications
Antigen | PCDHB2 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121699
![]() |
Thermo Scientific
PA5121699 |
100 μL |
Each for $405.00
|
|
PIPA5121700
![]() |
Thermo Scientific
PA5121700 |
20 μL |
Each for $143.00
|
|
Description
PCDHB2 Polyclonal antibody specifically detects PCDHB2 in Human, Mouse, Porcine, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
PCDHB2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Porcine, Rat | |
PCDHB2 | |
Q9Y5E7 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
PBS with 50% glycerol and 0.05% Proclin 300 | |
PCDHB2 | |
Primary | |
-20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title