Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCDHB5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5139885

This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DAGAYGSVAY ALFQGDEVTQ PFVIDEKTAE IRLKRALDFE ATPYYNVEIV
PCDHB5 Polyclonal antibody specifically detects PCDHB5 in Human samples. It is validated for ImmunocytochemistrySpecifications
PCDHB5 | |
Polyclonal | |
Unconjugated | |
PCDHB5 | |
cell-cell adhesion molecule; Pcdhb4A; PCDHB5; PcdhbE; PCDH-BETA5; PCDH-beta-5; protocadherin beta 5; protocadherin beta-5; Protocadherin-betaE | |
Recombinant protein corresponding to Human PCDHB5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry | |
0.1 μg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5E4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26167 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title