Learn More
PCDHGA1 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: S159-5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242207FR
Catalog No. NBP242207FR
Add to cart
Description
Description
PCDHGA1 Monoclonal antibody specifically detects PCDHGA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
PCDHGA1
S159-5
Western Blot, Immunohistochemistry
Purified
PCDHGA1
Fusion protein corresponding to amino acids 808-931 (C-terminal cytoplasmic constant domain) of mouse Protocadherin-gamma-A1 that is shared by all 22 Gamma-protocadherins (A subfamily amino acids ~807-930, B subfamily amino acids ~789-912 and C subfamily amino acids ~818-941). 99% identity with human (123/124 amino acids).
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 100kDa. Cross-reacts with all Gamma-protocadherin-A, -B and -C proteins.
Western Blot, Immunohistochemistry
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
MGC138287, MGC138289, PCDH-GAMMA-A1, protocadherin gamma subfamily A, 1, protocadherin gamma-A1
Mouse
IgG1
0.1 ml
Primary
56114
Human, Mouse, Rat