Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHGA3 Mouse anti-Human, Mouse, Clone: S144-17, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527616

Catalog No. PIMA527616


Add to cart

Description

Description

Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintece of specific neuronal connections in the brain.
Specifications

Specifications

PCDHGA3
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q9Y5H0
PCDHGA3, PCDG3, PCDH gamma A3, Protocadherin gamma subfamily A 3, Protocadherin gamma-A3, Gamma Protocadherin-A3, Gamma-Protocadherin-A3
Mouse
IgG1
100 μg
-20°C
Primary
56112, 93711
Immunocytochemistry, Immunofluorescence, Western Blot
S144-17
Unconjugated
PCDHGA3
Liquid
Pcdhga3
Fusion protein amino acids 720-804 (variable cytoplasmic domain) of mouse Gamma- protocadherin-A3. Human: 80% identity (68/84 amino acids identical) >75% identity with other Gamma-protocadherin-A protein.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit