Learn More
PCDHGA3 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: S144-32, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242184IR
Catalog No. NBP242184IR
Add to cart
Description
Description
PCDHGA3 Monoclonal antibody specifically detects PCDHGA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
PCDHGA3
S144-32
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PCDHGA3
Fusion protein amino acids 720-804 (variable cytoplasmic domain) of mouse Gamma-protocadherin-A3. Human: 80% identity (68/84 amino acids identical) >75% identity with other Gamma-protocadherin-A proteins
Protein G purified
Store at 4C in the dark.
Monoclonal
Geater then 75% identity with other Gamma-protocadherin-A proteinsDetects approx 100kDa. Cross-reacts with other Gamma protocadherin-A proteins. No cross-reactivity against Gamma protocadherin-B or -C proteins.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
PCDH-GAMMA-A3, protocadherin gamma subfamily A, 3, protocadherin gamma-A3
Mouse
IgG2b
0.1 ml
Primary
56112
Human, Mouse, Rat