Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHGA3 Mouse anti-Human, Mouse, FITC, Clone: S144-17, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242181F

Catalog No. NBP242181F


Add to cart

Description

Description

PCDHGA3 Monoclonal antibody specifically detects PCDHGA3 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

PCDHGA3
S144-17
Immunocytochemistry/Immunofluorescence
Purified
PCDHGA3
Fusion protein amino acids 720-804 (variable cytoplasmic domain) of mouse Gamma- protocadherin-A3. Human: 80% identity (68/84 amino acids identical) >75% identity with other Gamma-protocadherin-A protein.
Protein G purified
Store at 4C in the dark.
Monoclonal
Greater then 75% identity with other Gamma-protocadherin-A protein.Detects approx 100kDa. No cross-reactivity against other Protocadherin Gamma -A, -B or -C proteins.
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
PCDH-GAMMA-A3, protocadherin gamma subfamily A, 3, protocadherin gamma-A3
Mouse
IgG1
0.1 ml
Primary
56112
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit