Learn More
PCDHGA3 Mouse anti-Human, Mouse, HRP, Clone: S144-17, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242181H
Catalog No. NBP242181H
Add to cart
Description
Description
PCDHGA3 Monoclonal antibody specifically detects PCDHGA3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
PCDHGA3
S144-17
Western Blot
Purified
PCDHGA3
Fusion protein amino acids 720-804 (variable cytoplasmic domain) of mouse Gamma- protocadherin-A3. Human: 80% identity (68/84 amino acids identical) >75% identity with other Gamma-protocadherin-A protein.
Protein G purified
Store at 4C in the dark.
Monoclonal
Greater then 75% identity with other Gamma-protocadherin-A protein.Detects approx 100kDa. No cross-reactivity against other Protocadherin Gamma -A, -B or -C proteins.
Western Blot
HRP
PBS with No Preservative
PCDH-GAMMA-A3, protocadherin gamma subfamily A, 3, protocadherin gamma-A3
Mouse
IgG1
0.1 ml
Primary
56112
Human, Mouse