Learn More
Aviva Systems Biology Corp PCDHGA4 Antibody - N-terminal region (ARP49367_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49367_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCDHGA4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCDHGA4. Alternate names: PCDH-GAMMA-A4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-60074