Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCDHGB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567411
Description
Immunogen sequence: INAEITYAFLN SPISTSLFNL NPNTGDITTN GTLDFEETSR YVLSVEAKDG Highest antigen sequence identity to the following orthologs - mouse 79%, rat 75%.
Potential calcium-dependent cell-adhesion protein. May be involved in the establishment and maintenance of specific neuronal connections in the brain.
Specifications
PCDHGB1 | |
Polyclonal | |
Unconjugated | |
PCDHGB1 | |
PCDH-gamma-B1; PCDHGB1; protocadherin gamma subfamily B, 1; protocadherin gamma subfamily B, 1, isoform 2; protocadherin gamma-B1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56104 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y5G3 | |
PCDHGB1 | |
Recombinant Human PCDHGB1. Recombinant protein control fragment (Product #RP-108073). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction