Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCDHGB2 Mouse anti-Human, Mouse, Clone: S148-30, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527617

Catalog No. PIMA527617


Add to cart

Description

Description

This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Specifications

Specifications

PCDHGB2
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q9Y5G2
PCDH-gamma-B2, PCDHGB2, Protocadherin gamma B2, Protocadherin gamma subfamily B, 2, Protocadherin gamma subfamily B2, Gamma Protocadherin-B2, Gamma Protocadherin B2
Mouse
IgG1
100 μg
-20°C
Primary
56103, 93700
Immunocytochemistry, Immunofluorescence, Western Blot
S148-30
Unconjugated
PCDHGB2
Liquid
Pcdhgb2
Fusion protein amino acids 719-804 (variable cytoplasmic domain) of mouse Gamma- Protocadherin Gamma B2. Human: 68% identity (60/88 amino acids identical) >60% identity with other Gamma-protocadherin-B proteins
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit