Learn More
PCDHGB2 Mouse anti-Human, Mouse, DyLight 680, Clone: S148-30, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242183FR
Catalog No. NBP242183FR
Add to cart
Description
Description
PCDHGB2 Monoclonal antibody specifically detects PCDHGB2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
PCDHGB2
S148-30
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
PCDHGB2
Fusion protein amino acids 719-804 (variable cytoplasmic domain) of mouse Gamma- Protocadherin Gamma B2. Human: 68% identity (60/88 amino acids identical) >60% identity with other Gamma-protocadherin-B proteins
Protein G purified
Store at 4C in the dark.
Monoclonal
Greater then 60% identity with other Gamma-protocadherin-B proteins Detects approx 100kDa. No cross-reactivity against other Protocadherin Gamma -A, -B or -C proteins.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
MGC126854, PCDH-GAMMA-B2, protocadherin gamma subfamily B, 2, protocadherin gamma-B2
Mouse
IgG1
0.1 ml
Primary
56103
Human, Mouse