Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pcdhgb2, Mouse anti-Mouse, PerCP, Clone: S148-30, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB18562

Catalog No. 89-338-487


Add to cart

Description

Description

Specifications

Specifications

Pcdhgb2
S148-30
Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
protocadherin gamma subfamily B, 2
-
Mouse
IgG1
RUO
Antibody
Monoclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
PerCP
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide).
Liquid
Pcdhgb2
Recombinant protein corresponding to amino acids 719-804 at variable cytoplasmic domain of mouse Pcdhgb2.
100 μg
Store at -20°C.
Primary
93700
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit