Learn More
PCDHGB3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 212041AP
Catalog No. 50-173-3317
Add to cart
Specifications
Specifications
PCDHGB3
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y5G1
PCDH GAMMA B3, PCDHGB3, Protocadherin gamma B3
Rabbit
IgG
150 μL
-20°C
Primary
56102
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PCDHGB3
Liquid
PCDHGB3
PCDHGB3 Fusion Protein Ag15569
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse