Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pcdhgc3, Mouse anti-Mouse, ATTO565, Clone: S174B-27, Abnova
Mouse Monoclonal Antibody
Manufacturer: Abnova Corporation MAB18705
Specifications
Pcdhgc3 | |
S174B-27 | |
Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
protocadherin gamma subfamily C, 3 | |
MGC106426|PC43|Pcdh2 | |
Mouse | |
IgG1 | |
RUO | |
Antibody | |
Monoclonal | |
Human, Mouse |
Immunocytochemistry, Immunofluorescence, Western Blot | |
ATTO565 | |
In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
Liquid | |
Pcdhgc3 | |
Recombinant protein corresponding to amino acids 720-804 at variable cytoplasmic domain of mouse Pcdhgc3. | |
100 μg | |
Store at -20°C. | |
Primary | |
93706 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title