Learn More
PCF11 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303705A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Component of pre-mRNA cleavage complex II.Specifications
PCF11 | |
Polyclonal | |
Unconjugated | |
PCF11 | |
PCF11, cleavage and polyadenylation factor subunit, homolog S. cerevisiae | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51585 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
O94913 | |
PCF11 | |
Between 275 and 325 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.