Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCF11 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | PCF11 |
---|---|
Dilution | Immunohistochemistry |
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
PCF11 Polyclonal specifically detects PCF11 in Human samples. It is validated for Immunohistochemistry.Specifications
PCF11 | |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
KIAA0824PCF11p homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog, PCF11, cleavage and polyadenylation factor II subunit, homolog (S. cerevisiae), PCF11, cleavage and polyadenylation factor subunit, homolog (S. cerevisiae), pre-mRNA cleavage complex 2 protein Pcf11, Pre-mRNA cleavage complex II protein Pcf11 | |
The immunogen is a synthetic peptide directed towards the following sequence SSEHRLAGSRNKIINGIVQKQDTITEESEKQGTKPGRSSTRKRSRSRSPK (NP_056969). Peptide sequence SSEHRLAGSRNKIINGIVQKQDTITEESEKQGTKPGRSSTRKRSRSRSPK | |
Affinity purified |
Immunohistochemistry | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
51585 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title