Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCGF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549390
Description
Predicted to react with bovine based on sequence homology.
PCGF1 Polyclonal antibody specifically detects PCGF1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
PCGF1 | |
Polyclonal | |
Unconjugated | |
PCGF1 | |
2010002K04Rik; AU024121; nervous system polycomb 1; nervous system Polycomb-1; Nspc1; Pcgf1; polycomb group ring finger 1; polycomb group RING finger protein 1; RING finger protein 68; RNF3A-2; RNF68 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
312480, 69837, 84759 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q6DLV9, Q8R023, Q9BSM1 | |
PCGF1 | |
KLH conjugated synthetic peptide between 117-151 amino acids from the Central region of human PCGF1. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction