Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCIF1 Rabbit anti-Human, Polyclonal , Abnova™

PCIF1 polyclonal antibody raised against recombinant human PCIF1.

Manufacturer:  Abnova Corporation PAB29502

Catalog No. 89-934-737


Add to cart

Description

Description

Sequence: SSLVETPPAENKPRKRQLSEEQPSGNGVKKPKIEIPVTPTGQSVPSSPSIPGTPTLKMWGTSPEDKQQAALLRPTEVYWDLDIQTNAVIKHR
Specifications

Specifications

PCIF1
Unconjugated
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:500-1:1000) The optimal working dilution should be determined by the end user.
Liquid
PCIF1
Recombinant protein corresponding to amino acids of human PCIF1.
Antigen affinity purification
RUO
Primary
63935
Immunofluorescence, Immunohistochemistry (PFA fixed)
PCIF1 polyclonal antibody raised against recombinant human PCIF1.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
C20orf67
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit