Learn More
Aviva Systems Biology Corp PCK1 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP32800_P050
This is a rabbit polyclonal antibody against PCK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PCK1. Alternate names: MGC22652, PEPCK-C, PEPCK1, PEPCKCApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54770