Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCM1, Polyclonal, Bethyl Laboratories Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Bethyl Laboratories, IncSupplier Diversity Partner A301150AT

 View more versions of this product

Catalog No. 50-155-9384


Add to cart

Description

Description

The recommended shelf life for this product is 1 year from date of receipt.

PCM-1 is required for centrosome assembly and function. This protein is essential for the correct localization of several centrosomal proteins including CEP250, CETN3, PCNT and NEK2. The protein is required to anchor microtubules to the centrosome.
Specifications

Specifications

PCM1
Polyclonal
TBS with 0.1% BSA and 0.09% sodium azide
Q15154
Pericentriolar material 1 protein; PCM-1; hPCM-1 ; PCM1
Rabbit
IgG
20 μl
4° C
Primary
5108
Western Blot
Unconjugated
PCM1
liquid
PCM1
Between 1972 and 2022
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit