Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCMT1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297711APC
Description
PCMT1 Polyclonal antibody specifically detects PCMT1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
| PCMT1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry-Paraffin | |
| EC 2.1.1, EC 2.1.1.77, L-isoaspartyl protein carboxyl methyltransferase, L-isoaspartyl/D-aspartyl methyltransferase, PIMT, Protein L-isoaspartyl/D-aspartyl methyltransferase, Protein-beta-aspartate methyltransferase, protein-L-isoaspartate (D-aspartate) O-methyltransferase, protein-L-isoaspartate(D-aspartate) O-methyltransferase | |
| Produced in rabbits immunized with E. coli-derived Human PCMT1 fragment. | |
| 0.1 mL | |
| Proteases & Other Enzymes | |
| 5110 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction