Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCMTD1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP15437020UL

 View more versions of this product

Catalog No. NBP15437020


Add to cart

Description

Description

PCMTD1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

PCMTD1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
PCMTD1
Synthetic peptides corresponding to PCMTD1(protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1) The peptide sequence was selected from the middle region of PCMTD1. Peptide sequence TGQNTWESKNILAVSFAPLVQPSKNDNGKPDSV
41 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:100-1:2000
Q96MG8
FLJ10883, protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1, protein-L-isoaspartate O-methyltransferase domain-containing protein 1
Rabbit
IgG
Protein A purified
RUO
Primary
115294
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit