Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA associated factor Rabbit anti-Human, Alexa Fluor 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP299681AF488

Catalog No. NB062431


Add to cart

Description

Description

PCNA associated factor Polyclonal antibody specifically detects PCNA associated factor in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

PCNA associated factor
Alexa Fluor 488
50mM Sodium Borate
HCV NS5A-transactivated protein 9, Hepatitis C virus NS5A-transactivated protein 9, KIAA0101, NS5ATP9p15PAF, OEATC-1OEATC1, Overexpressed in anaplastic thyroid carcinoma 1, p15(PAF), PAF, PCNA-associated factor
Produced in rabbits immunized with purified, recombinant PCNA associated factor (NP_055551.1; Met1-Glu111).
Affinity purified
RUO
Store at 4°C in the dark.
Polyclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
Rabbit
IgG
0.1 mL
Cancer
Primary
9768
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit