Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCNA Mouse anti-Human, DyLight 550, Clone: 7H4F8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237414R

Catalog No. NBP237414R

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PCNA
7H4F8
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
PCNA
Purified recombinant fragment of human PCNA (AA: 53-196 ) expressed in E. Coli.
Protein A or G purified
Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers
Primary
5111
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit