Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

PCNA Mouse anti-Human, Insect, S. pombe, Alexa Fluor™ 594, Clone: PC5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB1002797AF594

Catalog No. N102797A594



Description

Description

PCNA Monoclonal antibody specifically detects PCNA in Human, Insect, S. pombe samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

PCNA
PC5
Western Blot, Immunohistochemistry
Conjugated, Purified
PCNA
Protein A-PCNA fusion obtained from pC2T.
Protein A or G purified
Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers
Primary
5111.0
Immunohistochemistry, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Insect, Yeast
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit