Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ PCNA Recombinant Human Protein, N-His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding mature form of human PCNA (Met 1-Ser 261) fused to polyhistidine tag at N-terminus

Manufacturer:  Sino Biological™ 12131H07B250

 View more versions of this product

Catalog No. 12-131-H07B25


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding mature form of human PCNA (Met 1-Ser 261) fused to polyhistidine tag at N-terminus

  • Formulation: 50mM Na3PO4, 300mM NaCl, 10% Glycerol, 2mM DTT, pH 7.0
Description & Specifications

Specifications

Analysis of protein stucture and used in cell biology research applications
5111
≥85% by SDS-PAGE
Human
<1 EU/μg
PCNA
Other Proteins
PCNA Recombinant Human Protein
Lyophilized
31kd
5 x 50μg
Full Length, Recombinant
Proliferating Cell Nuclear Antigen
Assay Standard
Insect
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit