Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCNP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570314
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
PCNP is a protein coding gene, and may be involved in cell cycle regulation.
Specifications
PCNP | |
Polyclonal | |
Unconjugated | |
PCNP | |
1110018D06Rik; Ab2-416; AI647035; Liver regeneration-related protein LRRG084; PCNP; PEST proteolytic signal containing nuclear protein; PEST proteolytic signal-containing nuclear protein; PEST-containing nuclear protein; Unknown (protein for MGC:128940); zgc:103440 | |
Rabbit | |
Affinity chromatography | |
RUO | |
76302 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6P8I4 | |
PCNP | |
synthetic peptide corresponding to a region of mouse PCNP (aa 112-161). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction