Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCNP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557113
Description
Immunogen sequence: NEDEDSEPEE MPPEAKMRMK NIGRDTPTSA GPNSFNKGKH GFSDNQKLWE RNIKSHLGNV HDQD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
PCNP is a protein coding gene, and may be involved in cell cycle regulation.
Specifications
PCNP | |
Polyclonal | |
Unconjugated | |
PCNP | |
1110018D06Rik; Ab2-416; AI647035; Liver regeneration-related protein LRRG084; PCNP; PEST proteolytic signal containing nuclear protein; PEST proteolytic signal-containing nuclear protein; PEST-containing nuclear protein; Unknown (protein for MGC:128940); zgc:103440 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57092 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WW12 | |
PCNP | |
Recombinant protein corresponding to Human PCNP. Recombinant protein control fragment (Product #RP-96291). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction