Learn More
Newcomer Supply Pneumocystis sp., Animal Control Slides, 98/set
SDP
Supplier:  Newcomer Supply 4556B
Encompass
Newcomer Supply Pneumocystis sp., Animal Control Slides are for the positive histochemical staining of fungal organisms. The morphology of the organisms is consistent with Pneumocystis sp. Pneumocystis sp., Animal Control Slides contain sections of positive staining primate lung and negative staining human lung.
Catalog No. NC0994761
Item Discontinued This item has been discontinued by Fisher Scientific. Please to view product availability in your area.