Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549455

Catalog No. PA549455


Add to cart

Description

Description

May function as a cell-type specific modulator for G protein-mediated cell signaling.
Specifications

Specifications

PCP2
Polyclonal
PBS with 0.09% sodium azide
Q8IVA1
Purkinje cell protein 2 homolog, PCP2
Rabbit
IgG
80 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
126006
Western Blot
Unconjugated
PCP2
Liquid
PCP2
KLH conjugated synthetic peptide between 91-125 amino acids from the Central region of human PCP2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit