Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCPE-1/PCOLCE Mouse anti-Human, Alexa Fluor™ 594, Clone: 18, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP223493AF594

Catalog No. NP223493A94


Add to cart

Description

Description

PCPE-1/PCOLCE Monoclonal antibody specifically detects PCPE-1/PCOLCE in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PCPE-1/PCOLCE
18
Western Blot, ELISA
Conjugated, Purified
PCOLCE
PICP purified from second trimester amniotic fluid by affinity chromatography
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Clone 18 is specific for for native PICP from amniotic fluid. Crossreactivity to other species is not determined.
ELISA, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
PCPE, PCPE-1, PCPE1Type I procollagen COOH-terminal proteinase enhancer, procollagen C-endopeptidase enhancer, procollagen C-endopeptidase enhancer 1, Procollagen COOH-terminal proteinase enhancer 1, Procollagen C-proteinase enhancer 1, procollagen, type 1, COOH-terminal proteinase enhancer, Type 1 procollagen C-proteinase enhancer protein
Mouse
IgG1 Kappa
0.1 mL
Primary
5118.0
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit