Thermo Scientific™ PCR Tubes and Plates Available on GSA/VA Contract for Federal Government customers only.

Efficiently perform applications with these PCR tubes and plates, molded from 100% virgin polypropylene.

Manufacturer: Thermo Scientific™ 3415

 View more versions of this product

Catalog No. 21-236-67A

  Check Availability
Add to Cart

    Specifications

    Cap Type Flat
    For Use With (Equipment) Thermal Cyclers
    Tube Type Plate
    Type Plates
    Wells 24
    Volume (Metric) 0.2mL
    Color Natural
    Description Thin-wall 24-well PCR Plates
    Item Description PCR Plate; 0.2mL; 24 well; Natural

    Efficiently perform applications with Thermo Scientific™ PCR Tubes and Plates, molded from 100% virgin polypropylene.

    • Tubes feature thin walls for optimal heat transfer
    • Caps seal securely to minimize evaporation.
    • Elevated well rims to ensure uniform sealing.
    • Thermo Scientific™ PurePak™ packaging reduces contamination
    • Certified RNase-, DNase-, DNA, and Pyrogen-free
    • Clear, nonsterile

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit