Learn More
Thermo Scientific™ PCR Tubes and Plates Available on GSA/VA Contract for Federal Government customers only.
Efficiently perform applications with these PCR tubes and plates, molded from 100% virgin polypropylene.
Manufacturer:  Thermo Scientific™ 3419
 View more versions of this product
Catalog No. 21-236-68A
  Check Availability
Add to cart
Specifications
Description
Efficiently perform applications with Thermo Scientific™ PCR Tubes and Plates, molded from 100% virgin polypropylene.
  • Tubes feature thin walls for optimal heat transfer
  • Caps seal securely to minimize evaporation.
  • Elevated well rims to ensure uniform sealing.
  • Thermo Scientific™ PurePak™ packaging reduces contamination
  • Certified RNase-, DNase-, DNA, and Pyrogen-free
  • Clear, nonsterile
Description & Specifications
Specifications
Plates
Natural
PCR Plate; 0.2mL; 48 well; Natural
0.2mL
Plate
Flat
Thin-wall 48-well PCR Plates
48
Thermal Cyclers