Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ PCR Tubes and Plates

Efficiently perform applications with these PCR tubes and plates, molded from 100% virgin polypropylene.

Manufacturer:  Thermo Scientific™ 3414

 View more versions of this product

Catalog No. 21-236-66

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Efficiently perform applications with Thermo Scientific™ PCR Tubes and Plates, molded from 100% virgin polypropylene.

  • Tubes feature thin walls for optimal heat transfer
  • Caps seal securely to minimize evaporation.
  • Elevated well rims to ensure uniform sealing.
  • Thermo Scientific™ PurePak™ packaging reduces contamination
  • Certified RNase-, DNase-, DNA, and Pyrogen-free
  • Clear, nonsterile
Specifications

Specifications

0.2 mL
Non-sterile
Domed
PCR thin wall tubes with dome caps
0.2 mL
Individual
Polypropylene
Tubes
Natural
PCR Tube; 0.2mL; Natural; With dome caps
Thermal Cyclers
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit