Learn More
Abnova™ PCSK1N 293T Cell Transient Overexpression Lysate (Denatured) (T02)
Human overexpression lysate
Manufacturer:  Abnova™ H00027344T02
Catalog No. 89-956-082
Add to cart
Specifications
Specifications
293T
NM_013271
pCMV-PCSK1N full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
PCSK1N 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 27.4kDa
Transient overexpression cell lysate was tested with anti-PCSK1N antibody (Mfr. No. H00027344-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human