Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCSK4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548804

Catalog No. PA548804

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Involved in the processing of hormone and other protein precursors at sites comprised of pairs of basic amino acid residues. Plays a role in transcriptional coactivation. May be involved in stabilizing the multiprotein transcription complex.
Specifications

Specifications

PCSK4
Polyclonal
PBS with 0.09% sodium azide
Q6UW60
Proprotein convertase subtilisin/kexin type 4, 3421-, Proprotein convertase 4, PC4, PCSK4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54760
Western Blot
Unconjugated
PCSK4
Liquid
PCSK4
KLH conjugated synthetic peptide between 168-196 amino acids from the N-terminal region of human PCSK4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit