Learn More
PCSK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151061AP
Catalog No. 50-172-9983
Add to cart
Description
Description
Involved in the processing of hormone and other protein precursors at sites comprised of pairs of basic amino acid residues. Plays a role in transcriptional coactivation. May be involved in stabilizing the multiprotein transcription complex.
Specifications
Specifications
PCSK4
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P29121, Q6UW60, Q78EH2
PC4, PCSK4, Proprotein convertase 4, SPC5
Rabbit
IgG
150 μL
-20°C
Primary
171085, 18551, 54760
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PCSK4
Liquid
Pcsk4
PCSK4 Fusion Protein Ag4007
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat