Learn More
Aviva Systems Biology Corp PCSK5 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP43530_P050
This is a rabbit polyclonal antibody against PCSK5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PCSK5. Alternate names: PC5, PC6, PC6A, SPC6ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53369