Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCTP Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548782

Catalog No. PA548782


Add to cart

Description

Description

Catalyzes the transfer of phosphatidylcholine between membranes. Binds a single lipid molecule.
Specifications

Specifications

PCTP
Polyclonal
PBS with 0.09% sodium azide
Q9UKL6
Phosphatidylcholine transfer protein, PC-TP, START domain-containing protein 2, StARD2, StAR-related lipid transfer protein 2, PCTP, STARD2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
58488
Western Blot
Unconjugated
PCTP
Liquid
PCTP
KLH conjugated synthetic peptide between 59-87 amino acids from the Central region of human PCTP.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit