Learn More
PD-1 Armenian Hamster anti-Mouse, Alexa Fluor 700, Clone: J43, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143110AF700
Catalog No. NBP143110AI
Add to cart
Description
Description
PD-1 Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications
Specifications
PD-1
J43
Flow Cytometry, Immunohistochemistry-Frozen
Purified
PDCD1
The immunogen for this antibody was PD1.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry (Frozen)
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Armenian Hamster
IgG
0.1 ml
Apoptosis, Stem Cell Markers
Primary
5133