Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (J43), Allophycocyanin, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP143110APC
Description
PD-1 Monoclonal specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
PD-1 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
PDCD1 | |
The immunogen for this antibody was PD1. | |
0.1 mL | |
Apoptosis, Stem Cell Markers | |
5133 | |
Store at 4C in the dark. | |
IgG |
Flow Cytometry | |
J43 | |
Flow Cytometry | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
Armenian Hamster | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction