Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Armenian Hamster anti-Mouse, PerCP, Clone: J43, Novus Biologicals™

Armenian Hamster Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143110PCP

Catalog No. NBP143110JP


Add to cart

Description

Description

PD-1 Monoclonal antibody specifically detects Antigen in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).
Specifications

Specifications

PD-1
J43
Flow Cytometry, Immunohistochemistry-Frozen
Purified
PDCD1
The immunogen for this antibody was PD1.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry (Frozen)
PerCP
PBS with 0.05% Sodium Azide
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Armenian Hamster
IgG
0.1 ml
Apoptosis, Stem Cell Markers
Primary
5133
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit