Learn More
PD-1 Mouse anti-Human, DyLight 755, Clone: 913425, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals MAB10862IR
Catalog No. MAB10862IR
Add to cart
Description
Description
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
PD-1
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
PDCD1
rhPD-1 aa25-167
100 μL
Apoptosis, Stem Cell Markers
5133
Store at 4C in the dark.
IgG1
Flow Cytometry
913425
Flow Cytometry
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Mouse
Protein A or G purified
RUO
Primary
Human
Purified