Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Mouse anti-Human, DyLight 550, Clone: J116, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143107R

Catalog No. NBP143107R


Add to cart

Description

Description

PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

PD-1
J116
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
PDCD1
The immunogen for this antibody was PD1.
Protein A or G purified
Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
Primary
5133.0
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Mouse
IgG1 Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit