Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Rabbit anti-Human, Alexa Fluor™ 594, Clone: 2335A, R&D Systems™

Rabbit Monoclonal Antibody

Manufacturer:  R&D Systems FAB10863T100UG

Catalog No. FB10863T100


Add to cart
This item is not returnable. View return policy

Description

Description

PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

PD-1
2335A
Flow Cytometry 0.25-1 μg/106 cells
Q15116
Rabbit
IgG
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human PD-1 in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 594
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Mouse myeloma cell line, NS0-derived human PD-1, Met1-Gln167, Accession No. Q15116
100 μg
Primary
5133
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit