Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Rabbit anti-Human, PE-Cyanine7, Clone: PDCD1/1410R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254570PECY7

Catalog No. N254570PEC7


Add to cart

Description

Description

PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

PD-1
PDCD1/1410R
Flow Cytometry
Conjugated, Purified
PDCD1
Human recombinant PDCD1 protein fragment (exact sequence is proprietary)
Protein A or G purified
Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
Primary
5133.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Rabbit
IgG
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit