Learn More
PD-1 Rat anti-Mouse, Alexa Fluor™ 750, Clone: 996221, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB77381S100UG
Catalog No. FAB77381S10
Add to cart
This item is not returnable. View return policy
Description
Description
PD-1 Monoclonal specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
PD-1
996221
Flow Cytometry 0.25-1 μg/106 cells
Q02242
CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Mouse myeloma cell line, NS0-derived mouse PD-1, Met1-Gln167, Accession No. Q02242
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Monoclonal
Detects mouse PD-1 in direct ELISAs.
Flow Cytometry
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Rat
IgG2a
100 μg
Primary
5133.0
Mouse